Anti-BAG1 Rabbit Polyclonal Antibody
BWRLBS6821
- Antibody type:Primary
- Antigen symbol:BAG1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human BAG1.
BAG1 pAb detects endogenous levels of BAG1 protein.
Type: Primary
Antigen: BAG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat