Anti-BMP2 Rabbit Polyclonal Antibody
Catalog # BWRLBS6815
Supplier: Bioworld Technology
Specifications
- Antistof type:Primary
- Antigen symbol:BMP2
- Clonality:Polyclonal
- Vært:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
Recombinant full length Human BMP2.
BMP2 pAb detects endogenous levels of BMP2 protein.
Type: Primary
Antigen: BMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat