Anti-p-Gab 1 Rabbit Polyclonal Antibody [clone: Y627]
Catalog # BWRLBS4790
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Gab 1
- Clonality:Polyclonal
- Clone:Y627
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Gab 1 around the phosphorylation site of Tyrosine 627.
p-Gab 1 (Y627) pAb detects endogenous levels of Gab 1 protein only when phosphorylated at Tyr627.
Type: Primary
Antigen: p-Gab 1
Clonality: Polyclonal
Clone: Y627
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat