Anti-Trk A Rabbit Polyclonal Antibody
Catalog # BWRLBS6895
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Trk A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human Trk A.
Trk A pAb detects endogenous levels of Trk A protein.
Type: Primary
Antigen: Trk A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human