Anti-Six3/6 Rabbit Polyclonal Antibody [clone: D228]
Catalog # BWRLBS1903
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Six3/6
- Clonality:Polyclonal
- Clone:D228
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 201-250 of Human Six3.
Six3/6 (D228) pAb detects endogenous levels of Six3/6 protein.
Type: Primary
Antigen: Six3/6
Clonality: Polyclonal
Clone: D228
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat