Anti-BAI-1 Rabbit Polyclonal Antibody [clone: Q732]
Catalog # BWRLBS2784
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:BAI-1
- Clonality:Polyclonal
- Clone:Q732
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 711-760 of Human BAI-1.
BAI-1 (Q732) pAb detects endogenous levels of BAI-1 protein.
Type: Primary
Antigen: BAI-1
Clonality: Polyclonal
Clone: Q732
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse