Anti-IKKgamma Rabbit Polyclonal Antibody [clone: H81]
Catalog # BWRLBS1843
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IKK gamma
- Clonality:Polyclonal
- Clone:H81
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human IKKgamma.
IKK? (H81) pAb detects endogenous levels of IKK? protein.
Type: Primary
Antigen: IKKgamma
Clonality: Polyclonal
Clone: H81
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat