Anti-AURKA Rabbit Polyclonal Antibody [clone: Y338]
Catalog # BWRLBS1840
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:AURKA
- Clonality:Polyclonal
- Clone:Y338
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 311-360 of Human AURKA.
AURKA (Y338) pAb detects endogenous levels of AURKA protein.
Type: Primary
Antigen: AURKA
Clonality: Polyclonal
Clone: Y338
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat