Anti-Smad2/3 Rabbit Polyclonal Antibody [clone: S2]
BWRLBS1838
- Antibody type:Primary
- Antigen symbol:Smad2/3
- Clonality:Polyclonal
- Clone:S2
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminal of Human Smad2.
Smad2/3 (S2) pAb detects endogenous levels of Smad2/3 protein.
Type: Primary
Antigen: Smad2/3
Clonality: Polyclonal
Clone: S2
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat