Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-Smad1 Rabbit Polyclonal Antibody [clone: P183]
Anti-Smad1 Rabbit Polyclonal Antibody [clone: P183]
  BWRLBS1837
Anti-Smad1 Rabbit Polyclonal Antibody [clone: P183]
  BWRLBS1837

 

  • Antistof type:
    Primary
  • Antigen symbol:
    SMAD family member 1
  • Clonality:
    Polyclonal
  • Clone:
    P183
  • Vært:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunFluorescens:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Størrelse:
    100 µg
  • Pakn:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 152-201 of Human Smad1.

Smad1 (P183) pAb detects endogenous levels of Smad1 protein.

Type: Primary
Antigen: Smad1
Clonality: Polyclonal
Clone: P183
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse