Anti-BRS-3 Rabbit Polyclonal Antibody [clone: C203]
Catalog # BWRLBS2769
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:BRS-3
- Clonality:Polyclonal
- Clone:C203
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 280-330 of Human BRS-3.
BRS-3 (C203) pAb detects endogenous levels of BRS-3 protein.
Type: Primary
Antigen: BRS-3
Clonality: Polyclonal
Clone: C203
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat