Order Entry
Denmark
ContactUsLinkComponent
 
Anti-14-3-3 zeta Rabbit Polyclonal Antibody [clone: W228]
  BWRLBS1826
Anti-14-3-3 zeta Rabbit Polyclonal Antibody [clone: W228]
  BWRLBS1826

 

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 zeta
  • Clonality:
    Polyclonal
  • Clone:
    W228
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 200-250 of Human 14-3-3 zeta.

14-3-3 ? (W228) pAb detects endogenous levels of 14-3-3 protein zeta/delta.

Type: Primary
Antigen: 14-3-3 zeta
Clonality: Polyclonal
Clone: W228
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat