Anti-IP Receptor Rabbit Polyclonal Antibody [clone: H237]
BWRLBS3196
- Antibody type:Primary
- Antigen symbol:IP Receptor
- Clonality:Polyclonal
- Clone:H237
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 200-250 of Human IP Receptor.
IP Receptor (H237) pAb detects endogenous levels of IP Receptor protein.
Type: Primary
Antigen: IP Receptor
Clonality: Polyclonal
Clone: H237
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human