Order Entry
Denmark
ContactUsLinkComponent
 
Anti-GPR18 Rabbit Polyclonal Antibody [clone: C172]
  BWRLBS3164
Anti-GPR18 Rabbit Polyclonal Antibody [clone: C172]
  BWRLBS3164

 

  • Antibody type:
    Primary
  • Antigen symbol:
    GPR18
  • Clonality:
    Polyclonal
  • Clone:
    C172
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 140-190 of Human GPR18.

GPR18 (C172) pAb detects endogenous levels of GPR18 protein.

Type: Primary
Antigen: GPR18
Clonality: Polyclonal
Clone: C172
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat