Order Entry
Denmark
ContactUsLinkComponent
Anti-EP3 Rabbit Polyclonal Antibody [clone: R39]
Anti-EP3 Rabbit Polyclonal Antibody [clone: R39]
Catalog # BWRLBS3162
Anti-EP3 Rabbit Polyclonal Antibody [clone: R39]
Catalog # BWRLBS3162

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    EP3
  • Clonality:
    Polyclonal
  • Clone:
    R39
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids N-terminus of Human EP3.

EP3 (R39) pAb detects endogenous levels of EP3 protein.

Type: Primary
Antigen: EP3
Clonality: Polyclonal
Clone: R39
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human