Anti-D4DR Rabbit Polyclonal Antibody [clone: L396]
BWRLBS3160
- Antibody type:Primary
- Antigen symbol:D4DR
- Clonality:Polyclonal
- Clone:L396
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 370-420 of Human D4DR.
D4DR (L396) pAb detects endogenous levels of D4DR protein.
Type: Primary
Antigen: D4DR
Clonality: Polyclonal
Clone: L396
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat