Anti-CCK-BR Rabbit Polyclonal Antibody [clone: L54]
Catalog # BWRLBS3159
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CCK-BR
- Clonality:Polyclonal
- Clone:L54
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 30-80 of Human CCK-BR.
CCK-BR (L54) pAb detects endogenous levels of CCK-BR protein.
Type: Primary
Antigen: CCK-BR
Clonality: Polyclonal
Clone: L54
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat