Ordreindtastning
Denmark
ContactUsLinkComponent
 
Anti-GPR150 Rabbit Polyclonal Antibody [clone: Q391]
  BWRLBS2702
Anti-GPR150 Rabbit Polyclonal Antibody [clone: Q391]
  BWRLBS2702

 

  • Antistof type:
    Primary
  • Antigen symbol:
    GPR150
  • Clonality:
    Polyclonal
  • Clone:
    Q391
  • Vært:
    Rabbit
  • ImmunFluorescens:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Størrelse:
    100 µg
  • Pakn:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 360-410 of Human GPR150.

GPR150 (Q391) pAb detects endogenous levels of GPR150 protein.

Type: Primary
Antigen: GPR150
Clonality: Polyclonal
Clone: Q391
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat