Anti-Rho H Rabbit Polyclonal Antibody [clone: M134]
Catalog # BWRLBS2660
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Rho H
- Clonality:Polyclonal
- Clone:M134
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 102-152 of Human Rho H.
Rho H (M134) pAb detects endogenous levels of Rho H protein.
Type: Primary
Antigen: Rho H
Clonality: Polyclonal
Clone: M134
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat