Anti-IGF-2R Rabbit Polyclonal Antibody [clone: F2480]
Catalog # BWRLBS1755
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IGF-2R
- Clonality:Polyclonal
- Clone:F2480
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 2441-2490 of Human IGF-2R.
IGF-2R (F2480) pAb detects endogenous levels of IGF-2R protein.
Type: Primary
Antigen: IGF-2R
Clonality: Polyclonal
Clone: F2480
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat