Anti-Histone H3.3 Rabbit Polyclonal Antibody [clone: K27]
Catalog # BWRLBS1752
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H3.3
- Clonality:Polyclonal
- Clone:K27
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human Histone H3.3.
Histone H3.3 (K27) pAb detects endogenous levels of Histone H3.3 protein.
Type: Primary
Antigen: Histone H3.3
Clonality: Polyclonal
Clone: K27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat