Anti-SRp20 Rabbit Polyclonal Antibody [clone: F151]
BWRLBS2559
- Antibody type:Primary
- Antigen symbol:SRp20
- Clonality:Polyclonal
- Clone:F151
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 120-170 of Human SRp20.
SRp20 (F151) pAb detects endogenous levels of SRp20 protein.
Type: Primary
Antigen: SRp20
Clonality: Polyclonal
Clone: F151
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse