Anti-DNA-PK Rabbit Polyclonal Antibody [clone: Y2052]
Catalog # BWRLBS1634
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:DNA-PK
- Clonality:Polyclonal
- Clone:Y2052
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 2022-2070 of Human DNA-PK.
DNA-PK (Y2052) pAb detects endogenous levels of DNA-PK protein.
Type: Primary
Antigen: DNA-PK
Clonality: Polyclonal
Clone: Y2052
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat