Anti-AURKA Rabbit Polyclonal Antibody [clone: P282]
Catalog # BWRLBS1612
Supplier: Bioworld Technology
Specifications
- Antistof type:Primary
- Antigen symbol:AURKA
- Clonality:Polyclonal
- Clone:P282
- Vært:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 251-300 of Human AURKA.
AURKA (P282) pAb detects endogenous levels of AURKA protein.
Type: Primary
Antigen: AURKA
Clonality: Polyclonal
Clone: P282
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat