Anti-NQO1 Rabbit Polyclonal Antibody [clone: F252]
Catalog # BWRLBS2945
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:NQO1
- Clonality:Polyclonal
- Clone:F252
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 220-370 of Human NQO1.
NQO1 (F252) pAb detects endogenous levels of NQO1 protein.
Type: Primary
Antigen: NQO1
Clonality: Polyclonal
Clone: F252
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human