Anti-NFAT4 Rabbit Polyclonal Antibody [clone: E159]
Catalog # BWRLBS1559
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:NFAT4
- Clonality:Polyclonal
- Clone:E159
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 130-180 of Human NFAT4.
NFAT4 (E159) pAb detects endogenous levels of NFAT4 protein.
Type: Primary
Antigen: NFAT4
Clonality: Polyclonal
Clone: E159
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse