Order Entry
Denmark
ContactUsLinkComponent
 
Anti-TAF I p48 Rabbit Polyclonal Antibody [clone: R318]
  BWRLBS2313
Anti-TAF I p48 Rabbit Polyclonal Antibody [clone: R318]
  BWRLBS2313

 

  • Antibody type:
    Primary
  • Antigen symbol:
    TAF I p48
  • Clonality:
    Polyclonal
  • Clone:
    R318
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 280-336 of Human TAF I p48.

TAF I p48 (R318) pAb detects endogenous levels of TAF I p48 protein.

Type: Primary
Antigen: TAF I p48
Clonality: Polyclonal
Clone: R318
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat