Order Entry
Denmark
ContactUsLinkComponent
Anti-Vimentin Rabbit Polyclonal Antibody [clone: I444]
Anti-Vimentin Rabbit Polyclonal Antibody [clone: I444]
Catalog # BWRLBS1491
Anti-Vimentin Rabbit Polyclonal Antibody [clone: I444]
Catalog # BWRLBS1491

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Vimentin
  • Clonality:
    Polyclonal
  • Clone:
    I444
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 411-460 of Human Vimentin.

Vimentin (I444) pAb detects endogenous levels of Vimentin protein.

Type: Primary
Antigen: Vimentin
Clonality: Polyclonal
Clone: I444
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat