Anti-p-EGFR Rabbit Polyclonal Antibody [clone: S1046]
Catalog # BWRLBS4061
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-EGFR
- Clonality:Polyclonal
- Clone:S1046
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human EGFR around the phosphorylation site of Serine 1046.
p-EGFR (S1046) pAb detects endogenous levels of EGFR protein only when phosphorylated at serine 1046 (removal of the signal peptide).
Type: Primary
Antigen: p-EGFR
Clonality: Polyclonal
Clone: S1046
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat