Order Entry
Denmark
ContactUsLinkComponent
Anti-p-Cyclin E1 Rabbit Polyclonal Antibody [clone: T395]
Anti-p-Cyclin E1 Rabbit Polyclonal Antibody [clone: T395]
Catalog # BWRLBS4057
Anti-p-Cyclin E1 Rabbit Polyclonal Antibody [clone: T395]
Catalog # BWRLBS4057

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-Cyclin E1
  • Clonality:
    Polyclonal
  • Clone:
    T395
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human Cyclin E1 around the phosphorylation site of Threonine 395.

p-Cyclin E1 (T395) pAb detects endogenous levels of Cyclin E1 protein only when phosphorylated at Thr395

Type: Primary
Antigen: p-Cyclin E1
Clonality: Polyclonal
Clone: T395
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse