Order Entry
Denmark
ContactUsLinkComponent
Anti-p-CrkL Rabbit Polyclonal Antibody [clone: Y207]
Anti-p-CrkL Rabbit Polyclonal Antibody [clone: Y207]
Catalog # BWRLBS4056
Anti-p-CrkL Rabbit Polyclonal Antibody [clone: Y207]
Catalog # BWRLBS4056

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-CrkL
  • Clonality:
    Polyclonal
  • Clone:
    Y207
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human CrkL around the phosphorylation site of Tyrosine 207.

p-Crk-L (Y207) pAb detects endogenous levels of Crk-L protein only when phosphorylated at Tyr207.

Type: Primary
Antigen: p-CrkL
Clonality: Polyclonal
Clone: Y207
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat