Anti-p-CREB Rabbit Polyclonal Antibody [clone: S133]
Catalog # BWRLBS4053
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-CREB
- Clonality:Polyclonal
- Clone:S133
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human CREB around the phosphorylation site of Serine 133.
p-CREB (S133) pAb detects endogenous levels of CREB protein when phosphorylated at Ser133.
Type: Primary
Antigen: p-CREB
Clonality: Polyclonal
Clone: S133
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat