Order Entry
Denmark
ContactUsLinkComponent
Anti-p-c-Jun Rabbit Polyclonal Antibody [clone: Y170]
Anti-p-c-Jun Rabbit Polyclonal Antibody [clone: Y170]
Catalog # BWRLBS4050
Anti-p-c-Jun Rabbit Polyclonal Antibody [clone: Y170]
Catalog # BWRLBS4050

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-c-Jun
  • Clonality:
    Polyclonal
  • Clone:
    Y170
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human c-Jun around the phosphorylation site of Tyrosine 170.

p-c-Jun (Y170) pAb detects endogenous levels of c-Jun protein only when phosphorylated at Tyr170.

Type: Primary
Antigen: p-c-Jun
Clonality: Polyclonal
Clone: Y170
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat