Anti-IkB-alpha Rabbit Polyclonal Antibody [clone: S36]
Catalog # BWRLBS3601
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IkB-alpha
- Clonality:Polyclonal
- Clone:S36
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human IkB-alpha.
I?B-? (S36) pAb detects endogenous levels of I?B-? protein.
Type: Primary
Antigen: IkB-alpha
Clonality: Polyclonal
Clone: S36
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat