Anti-ABL1 Rabbit Polyclonal Antibody [clone: L406]
Catalog # BWRLBS3592
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ABL1
- Clonality:Polyclonal
- Clone:L406
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 381-430 of Human ABL1.
ABL1 (L406) pAb detects endogenous levels of ABL1 protein.
Type: Primary
Antigen: ABL1
Clonality: Polyclonal
Clone: L406
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat