Anti-FoxO3A Rabbit Polyclonal Antibody [clone: P247]
Catalog # BWRLBS3574
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:FoxO3A
- Clonality:Polyclonal
- Clone:P247
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 221-270 of Human FoxO3A.
FoxO3A (P247) pAb detects endogenous levels of FoxO3A protein.
Type: Primary
Antigen: FoxO3A
Clonality: Polyclonal
Clone: P247
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat