Order Entry
Denmark
ContactUsLinkComponent
 
Anti-NF-1 Rabbit Polyclonal Antibody [clone: Q34]
  BWRLBS2042
Anti-NF-1 Rabbit Polyclonal Antibody [clone: Q34]
  BWRLBS2042

 

  • Antibody type:
    Primary
  • Antigen symbol:
    NF-1
  • Clonality:
    Polyclonal
  • Clone:
    Q34
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to the N-terminus of Human NF-1.

NF-1 (Q34) pAb detects endogenous levels of NF-1 protein.

Type: Primary
Antigen: NF-1
Clonality: Polyclonal
Clone: Q34
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat