Anti-Casein Kinase Igamma2 Rabbit Polyclonal Antibody [clone: G7]
BWRLBS2025
- Antibody type:Primary
- Antigen symbol:Casein Kinase I gamma2
- Clonality:Polyclonal
- Clone:G7
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminal of Human Casein Kinase Igamma2.
Casein Kinase I?2 (G7) pAb detects endogenous levels of Casein Kinase I?2 protein.
Type: Primary
Antigen: Casein Kinase Igamma2
Clonality: Polyclonal
Clone: G7
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat