Anti-Peroxin 7 Rabbit Polyclonal Antibody [clone: F236]
BWRLBS3899
- Antibody type:Primary
- Antigen symbol:Peroxin 7
- Clonality:Polyclonal
- Clone:F236
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 211-260 of Human Peroxin 7.
Peroxin 7 (F236) pAb detects endogenous levels of Peroxin 7 protein.
Type: Primary
Antigen: Peroxin 7
Clonality: Polyclonal
Clone: F236
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat