Anti-T-type Ca++ CP alpha1H Rabbit Polyclonal Antibody [clone: P492]
Catalog # BWRLBS3413
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:T-type Ca++ CP alpha1H
- Clonality:Polyclonal
- Clone:P492
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 461-510 of Human T-type Ca++ CP alpha1H.
T-type Ca++ CP ?1H (P492) pAb detects endogenous levels of T-type Ca++ CP ?1H protein.
Type: Primary
Antigen: T-type Ca++ CP alpha1H
Clonality: Polyclonal
Clone: P492
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat