Anti-Nibrin Rabbit Polyclonal Antibody [clone: T337]
BWRLBS1282
- Antibody type:Primary
- Antigen symbol:Nibrin
- Clonality:Polyclonal
- Clone:T337
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 311-360 of Human Nibrin.
Nibrin (T337) pAb detects endogenous levels of Nibrin protein.
Type: Primary
Antigen: Nibrin
Clonality: Polyclonal
Clone: T337
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat