Anti-p70 S6K Rabbit Polyclonal Antibody [clone: F415]
Catalog # BWRLBS1280
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p70 S6K
- Clonality:Polyclonal
- Clone:F415
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 380-430 of Human p70 S6K.
p70 S6K (F415) pAb detects endogenous levels of p70 S6 Kinase protein.
Type: Primary
Antigen: p70 S6K
Clonality: Polyclonal
Clone: F415
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat