Anti-NSE/Enolase Rabbit Polyclonal Antibody [clone: R400]
Catalog # BWRLBS1259
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:NSE/Enolase
- Clonality:Polyclonal
- Clone:R400
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 361-410 of Human NSE/Enolase.
NSE/ENO2 (R400) pAb detects endogenous levels of NSE/ENO2 protein.
Type: Primary
Antigen: NSE/Enolase
Clonality: Polyclonal
Clone: R400
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat