Order Entry
Denmark
ContactUsLinkComponent
Superoxide dismutase assay kit
Superoxide dismutase assay kit
Catalog # ABNOKA3782
Supplier:  Abnova
CAS Number:  
undefined
Superoxide dismutase assay kit
Catalog # ABNOKA3782
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Superoxide Dismutase Assay Kit, 100 assays

Specifications

About this item

Superoxide Dismutase (SOD) Assay Kit is used for the determination of SOD in blood, cells, tissues and other biological samples.

  • Sensitive and accurate
  • Linear detection range of 0,05 to 3 U/ml SOD
  • Convenient and high-throughput
  • Store kit at −20 °C

This homogeneous mix-incubate-measure assay has no wash or reagent transfer steps involved. It can be readily automated on HTS liquid handling systems.

Delivery: Kit includes assay buffer (20 ml), SOD enzyme (120 µl), xanthine (600 µl), diluent (20 ml), XO enzyme (120 µl) and WST-1 (600 µl).