β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) ≥98%
:53-84-9
:
:C₂₁H₂₇N₇O₁₄P₂
:MFCD00036253
:663.43 g/mol
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
: Merck
Dichloromethane 99.8% stabilised for HPLC
: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
: Thermo Fisher Scientific
1-Butanol NF
: Spectrum Chemical
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
: PanReac AppliChem
Acetone, meets analytical specification of Ph.Eur.
: Thermo Fisher Scientific
Chloroform 99.8% stabilised for spectroscopy
: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
: Honeywell Chemicals
Xylene (mixture of isomers), J.T.Baker®
: Avantor