Electronic standard thermometers
: Amarell
Handy and precise universal thermometer with many fields of application.
- 'HOLD' on set-up
- Automatic power-off after 10 minutes
- Response time: T₉₀= 8 seconds
Probes are fast response stainless steel semiconductors (Lר: 125×3,5 mm).
Frequently Bought Together