Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Accessories for photometers monoparameter
Accessories for photometers monoparameter
Accessories for photometers monoparameter
Catalog # PALIPTC027
Supplier:
Palintest
CAS Number:
Expand image
Accessories for photometers monoparameter
Catalog # PALIPTC027
Supplier:
Palintest
CAS Number:
Specifications
Pk:
1
Description:
NDF validation standards for Chlorometer Duo
Specifications
About this item
Product Details
Product Details