Specifications
- Pk:1 KIT
- Host:
- Description:VECTASTAIN® Elite® ABC Kit (Standard)
- Environmentally Preferable:
Specifications
About this item
The VECTASTAIN® Elite® ABC kit is supplied without biotinylated secondary antibodies or blocking serum. The 'Standard' kit consists of Elite® A and B reagents, which can be used with any biotinylated primary or secondary antibody, biotinylated nucleic acid probe or other biotinylated molecule.
- Suitable for use on frozen or paraffin-embedded tissues, cell smears, or cytospin preparations
- Offers low background and amplified signal compared to conventional covalent conjugates
- Stock solutions packaged in convenient dropper bottles and each kit includes mixing bottles to prepare the working solutions
- Detailed instructions for use included
Ordering information: Each kit contains sufficient reagents to prepare 110 ml of each working solution (500 to 1000 sections).