Bottles, Wide Neck, with Screw Cap
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
Clear glass wide mouth bottles, with black PP cap. Ideal for sampling, storing and transporting powders and pastes.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- Round thread in accordance with DIN 168
Frequently Bought Together