HPLC frits
: Restek
These 6 mm frits are manufactured from fine, chromatographic grade 316 stainless steel and different sizes are available to fit most column and pore sizes. Compatible frit have to be chosen by checking its pore size with particle size of the packaging in the column.
Frequently Bought Together